Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glt28d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177130.3 | NP_796104.2 | 2 | 636 | ||
GenBank mRNA | AK029059.1 | - | 2 | 512 | ||
AK030588.1 | - | 2 | 525 | |||
BC121823.1 | - | 2 | 364 | 40107196 |