Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Heatr5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177171.4 | NP_796145.2 | 33 | 5612 | ||
GenBank mRNA | AK122493.1 | - | 22 | 3616 | ||
AK161444.1 | - | 22 | 3930 | |||
AK161523.1 | - | 5 | 972 | |||
BC063265.1 | - | 9 | 1625 | |||
BC086656.1 | - | 32 | 5437 | 30544145 |