Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sycp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177191.3 | NP_796165.2 | 8, 9 | 737 | ||
GenBank mRNA | AK014411.1 | - | 8, 9 | 765 | ||
DQ103262.1 | - | 8, 9 | 737 |