Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Islr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161535.1 | NP_001155007.1 | 2 | 2651 | ||
NM_001161536.1 | NP_001155008.1 | 4 | 2832 | |||
NM_001161537.1 | NP_001155009.1 | 3 | 2823 | |||
NM_001161538.1 | NP_001155010.1 | 4 | 2600 | |||
NM_001161539.1 | NP_001155011.1 | 4 | 2562 | |||
NM_001161540.1 | NP_001155012.1 | 3 | 2518 | |||
NM_001161541.1 | NP_001155013.1 | 3 | 2409 | |||
NM_177193.5 | NP_796167.2 | 3 | 2642 | |||
GenBank mRNA | AK043849.1 | - | 1 | 476 | ||
AK047348.1 | - | 1 | 2130 | |||
AK129367.1 | - | 3 | 2328 | |||
BC059068.1 | - | 3 | 2495 | 6825665 | ||
BC096531.1 | - | 2 | 2651 | 6826287 | ||
EU497918.1 | - | 3 | 2459 |