Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abi3bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014399.2 | NP_001014421.2 | 26 | 2462 | ||
NM_001014422.2 | NP_001014422.1 | 34 | 3032 | |||
NM_001014423.2 | NP_001014423.1 | 38 | 3272 | |||
NM_001014424.2 | NP_001014424.1 | 35 | 3080 | |||
NM_178790.4 | NP_848905.2 | 33 | 2972 | |||
GenBank mRNA | AB190339.1 | - | 33 | 2918 | ||
AB190340.1 | - | 26 | 2408 | |||
AB190341.1 | - | 34 | 2978 | |||
AB190342.1 | - | 38 | 3218 | |||
AB190343.1 | - | 35 | 3026 | |||
BC026627.1 | - | 11 | 981 | |||
BC145388.1 | - | 25 | 2350 | 9053946 | ||
BC150763.1 | - | 25 | 2350 | 9087674 |