Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abi3bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014399.2 | NP_001014421.2 | 7, 8 | 800 | ||
NM_001014422.2 | NP_001014422.1 | 7, 8 | 800 | |||
NM_001014423.2 | NP_001014423.1 | 7, 8 | 800 | |||
NM_001014424.2 | NP_001014424.1 | 7, 8 | 800 | |||
NM_178790.4 | NP_848905.2 | 7, 8 | 800 | |||
GenBank mRNA | AB190339.1 | - | 7, 8 | 746 | ||
AB190340.1 | - | 7, 8 | 746 | |||
AB190341.1 | - | 7, 8 | 746 | |||
AB190342.1 | - | 7, 8 | 746 | |||
AB190343.1 | - | 7, 8 | 746 | |||
AK043879.1 | - | 7, 8 | 774 | |||
AK051214.1 | - | 7, 8 | 770 | |||
AK086582.1 | - | 7, 8 | 803 | |||
BC145388.1 | - | 7, 8 | 763 | 9053946 | ||
BC150763.1 | - | 7, 8 | 763 | 9087674 |