Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem154 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177260.2 | NP_796234.1 | 6, 7 | 692 | ||
GenBank mRNA | AK037111.1 | - | 6, 7 | 604 | ||
AK081441.1 | - | 6, 7 | 692 | |||
AK154236.1 | - | 6, 7 | 607 | |||
AK170385.1 | - | 6, 7 | 560 | |||
AK170626.1 | - | 6, 7 | 507 | |||
AK170665.1 | - | 6, 7 | 540 | |||
BC096432.1 | - | 6, 7 | 575 | 5390573 | ||
BC113174.1 | - | 6, 7 | 632 | 40060792 | ||
BC113175.1 | - | 6, 7 | 632 | 40060793 |