Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177268.4 | NP_796242.2 | 1 | 1065 | ||
GenBank mRNA | AK052460.1 | - | 1 | 241 | ||
AK085853.1 | - | 1 | 246 | |||
AK140039.1 | - | 1 | 292 | |||
BC021802.1 | - | 1 | 254 | |||
BC037442.1 | - | 1 | 295 | |||
BC116229.1 | - | 1 | 234 | 40091581 | ||
BC116230.1 | - | 1 | 241 | 40091582 |