Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177268.4 | NP_796242.2 | 2 | 1165 | ||
GenBank mRNA | AK052460.1 | - | 2 | 341 | ||
AK085853.1 | - | 2 | 346 | |||
AK140039.1 | - | 2 | 392 | |||
BC021802.1 | - | 2 | 354 | |||
BC037442.1 | - | 2 | 395 | |||
BC116229.1 | - | 2 | 334 | 40091581 | ||
BC116230.1 | - | 2 | 341 | 40091582 |