Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slfn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183201.4 | NP_899024.3 | 3 | 761 | ||
GenBank mRNA | AK036486.1 | - | 3 | 762 | ||
AK039815.1 | - | 3 | 750 | |||
AK040359.1 | - | 3 | 748 | |||
AK171886.1 | - | 2 | 703 | |||
AY261804.1 | - | 2 | 723 | |||
AY261805.1 | - | 2 | 718 | |||
BC117909.1 | - | 2 | 641 | 40097236 |