Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177801.3 | NP_808469.1 | 4 | 624 | ||
GenBank mRNA | AK077068.1 | - | 4 | 625 | ||
BC117072.1 | - | 4 | 624 | 40126391 | ||
BC120775.1 | - | 4 | 623 | 40129698 | ||
DQ118647.1 | - | 4 | 610 |