Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam179b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177805.3 | NP_808473.2 | 9 | 3709 | ||
GenBank mRNA | AK084470.1 | - | 6 | 821 | ||
AK172948.1 | - | 9 | 3637 | |||
BC157958.1 | - | 9 | 3511 | 9088131 | ||
BC158008.1 | - | 10 | 3661 | 9007316 | ||
BC172117.1 | - | 9 | 3511 | 9054403 |