Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt61a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099792.1 | NP_001093262.1 | 2 | 539 | ||
NM_001099793.1 | NP_001093263.1 | 2 | 400 | |||
NM_177374.4 | NP_796348.2 | 1 | 841 | |||
GenBank mRNA | AK032824.1 | - | 1 | 804 | ||
AK049138.1 | - | 1 | 812 | |||
AK133579.1 | - | 1 | 815 | |||
BC051186.1 | - | 1 | 795 | 6468754 |