Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198570.1 | NP_001185499.1 | 6 | 872 | ||
NM_001198571.1 | NP_001185500.1 | 6 | 872 | |||
NM_198127.2 | NP_937760.1 | 5 | 854 | |||
GenBank mRNA | BC056345.1 | - | 5 | 675 | 5705048 | |
BC079545.1 | - | 6 | 757 | 6851145 | ||
BC079646.1 | - | 6 | 796 | 6850720 |