Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181546.3 | NP_853524.1 | 7, 8 | 1498 | ||
GenBank mRNA | AB102947.1 | - | 7, 8 | 1421 |