Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcmt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177846.3 | NP_808514.2 | 1 | 1287 | ||
GenBank mRNA | AK038626.1 | - | 968 | |||
AK054392.1 | - | 1 | 977 | |||
AK084642.1 | - | 1 | 1254 | |||
AK172552.1 | - | 1 | 968 | |||
AK172970.1 | - | 1 | 1242 | |||
BC071272.1 | - | 1 | 1230 |