Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_184088.1 | NP_908976.1 | 16 | 2549 | ||
GenBank mRNA | AK155220.1 | - | 16 | 2550 | ||
BC146602.1 | - | 15 | 2174 | 40099465 |