Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_184088.1 | NP_908976.1 | 22 | 3663 | ||
GenBank mRNA | AK155220.1 | - | 23 | 3811 | ||
BC040249.1 | - | 7 | 1070 | |||
BC040798.1 | - | 7 | 1070 | |||
BC146602.1 | - | 21 | 3288 | 40099465 |