Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxcl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203320.3 | NP_976065.1 | 3 | 330 | ||
GenBank mRNA | AK144158.1 | - | 3 | 332 | ||
AK150314.1 | - | 3 | 329 | |||
AK154334.1 | - | 3 | 329 | |||
AK155702.1 | - | 539 | ||||
AY311403.1 | - | 3 | 328 | |||
BC117014.1 | - | 3 | 285 | 40126333 | ||
BC117016.1 | - | 3 | 285 | 40126335 |