Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252519.1 | NP_001239448.1 | 6 | 1696 | ||
NM_177889.5 | NP_808557.2 | 7 | 1786 | |||
GenBank mRNA | AB010308.1 | - | 1 | 61 | ||
AK129467.1 | - | 2 | 3197 | |||
AK137630.1 | - | 1410 | ||||
BC025629.1 | - | 1 | 942 | |||
BC060530.1 | - | 6 | 1713 | 5684819 | ||
BX525285.1 | - | 1 | 89 |