Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177893.3 | NP_808561.2 | 4 | 1911 | ||
GenBank mRNA | AK173087.1 | - | 3 | 1369 |