Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmppb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177910.3 | NP_808578.1 | 5 | 724 | ||
GenBank mRNA | AK088295.1 | - | 5 | 706 | ||
AK148125.1 | - | 724 | ||||
AK158470.1 | - | 5 | 724 | |||
AK165816.1 | - | 5 | 726 | |||
AK188677.1 | - | 1 | 35 | |||
BC061207.1 | - | 6 | 598 | 30249330 |