Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177922.2 | NP_808590.1 | 8 | 844 | ||
GenBank mRNA | AK153712.1 | - | 2 | 269 | ||
BC048082.1 | - | 8 | 844 | 6312082 | ||
BC053920.1 | - | 2 | 605 |