Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045528.1 | NP_001038993.1 | 19 | 1981 | ||
GenBank mRNA | AK049459.1 | - | 19 | 1926 | ||
AK049509.1 | - | 19 | 1981 | |||
AK082565.1 | - | 19 | 1930 | |||
AK085087.1 | - | 19 | 1972 | |||
AK122343.1 | - | 12 | 1220 | |||
AK161705.1 | - | 19 | 1911 | |||
BC034098.1 | - | 9 | 918 | |||
BC060651.1 | - | 19 | 1895 | |||
BC137706.1 | - | 19 | 1915 | 8860725 |