Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045528.1 | NP_001038993.1 | 2 | 236 | ||
GenBank mRNA | AK049459.1 | - | 2 | 181 | ||
AK049476.1 | - | 2 | 157 | |||
AK049509.1 | - | 2 | 237 | |||
AK082565.1 | - | 2 | 183 | |||
AK085087.1 | - | 2 | 224 | |||
AK161169.1 | - | 2 | 132 | |||
AK161705.1 | - | 2 | 166 | |||
BC060651.1 | - | 2 | 150 | |||
BC137706.1 | - | 2 | 170 | 8860725 |