Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039466.1 | NP_001034555.1 | 2 | 323 | ||
NM_183037.2 | NP_898858.1 | 2 | 426 | |||
GenBank mRNA | AK132368.1 | - | 2 | 292 | ||
AK139421.1 | - | 1 | 290 | |||
AK170904.1 | - | 2 | 324 | |||
AY251388.1 | - | 2 | 306 | |||
BC065049.1 | - | 2 | 469 | 6418851 | ||
BC082299.1 | - | 2 | 426 | |||
CA315205.1 | - | 2 | 556 | |||
CA320885.1 | - | 2 | 426 |