Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zranb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207302.1 | NP_997185.1 | 7 | 1599 | ||
GenBank mRNA | BC145520.1 | - | 4 | 1212 |