Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290837.1 | NP_001277766.1 | 3 | 422 | ||
NM_011354.3 | NP_035484.1 | 3 | 399 | |||
GenBank mRNA | AA423654.1 | - | 3 | 338 | ||
AF073297.1 | - | 3 | 180 | |||
AK002351.1 | - | 3 | 175 | |||
AK008802.1 | - | 3 | 188 | |||
AK010749.1 | - | 3 | 231 | |||
AK012395.1 | - | 3 | 192 | |||
AK012603.1 | - | 3 | 178 | |||
AK018746.1 | - | 3 | 175 | |||
AK132746.1 | - | 2 | 829 | |||
AK151362.1 | - | 3 | 192 | |||
AK168858.1 | - | 3 | 179 | |||
AK169106.1 | - | 3 | 181 | |||
BC081455.1 | - | 3 | 183 | |||
BC092136.1 | - | 3 | 212 | 6814342 | ||
BY070249.1 | - | 3 | 238 |
Gene Symbol: Fam221b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK038644.1 | - | 3 | 2885 |