Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnjl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045530.2 | NP_001038995.1 | 4, 5 | 831 | ||
GenBank mRNA | AK143205.1 | - | 4, 5 | 797 | ||
BC139344.1 | - | 3, 4 | 592 | 8862366 | ||
BC139345.1 | - | 3, 4 | 592 | 8862367 |