Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm884 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033434.2 | NP_001028606.2 | 8, 9 | 2429 | ||
GenBank mRNA | AK029868.1 | - | 8, 9 | 2429 | ||
JQ790768.1 | - | 8, 9 | 739 | |||
JQ790769.1 | - | 8, 9 | 739 | |||
JQ790771.1 | - | 8, 9 | 739 | |||
JQ790772.1 | - | 8, 9 | 739 | |||
JQ790774.1 | - | 8, 9 | 739 | |||
JQ790775.1 | - | 8, 9 | 739 | |||
JQ790776.1 | - | 8, 9 | 739 | |||
JQ790777.1 | - | 8, 9 | 739 | |||
JQ790778.1 | - | 8, 9 | 739 | |||
JQ790779.1 | - | 6, 7 | 592 | |||
JQ790780.1 | - | 6, 7 | 592 | |||
JQ790781.1 | - | 6, 7 | 592 | |||
JQ790782.1 | - | 6, 7 | 592 | |||
JQ790785.1 | - | 7, 8 | 613 | |||
JQ790786.1 | - | 6, 7 | 595 | |||
JQ790787.1 | - | 6, 7 | 595 |