Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ighg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK134155.1 | - | 2 | 861 | ||
AK156420.1 | - | 3 | 554 | |||
BC010327.1 | - | 4 | 638 | 2651776 | ||
BC025447.1 | - | 4 | 640 | 5068622 | ||
BC092049.1 | - | 5 | 631 | 3661511 | ||
BC092269.1 | - | 4 | 620 | 4015795 | ||
V00799.1 | - | 1 | 173 | |||
X67210.1 | - | 4 | 619 |
Gene Symbol: Igh-V7183 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | D29667.1 | - | 4 | 482 |