Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nckap5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001884.1 | NP_001001884.1 | 10 | 3473 | ||
GenBank mRNA | AK048721.1 | - | 3 | 1003 | ||
BC072572.1 | - | 10 | 3473 | 5719876 |