Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201362.2 | NP_958750.1 | 11 | 1045 | ||
GenBank mRNA | AK078964.1 | - | 11 | 1047 | ||
BC046343.1 | - | 8 | 752 | 6487853 |