Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atraid | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027855.4 | NP_082131.2 | 3, 4 | 296 | ||
NM_212470.3 | NP_997635.1 | 3, 4 | 199 | |||
GenBank mRNA | AB009018.1 | - | 3, 4 | 279 | ||
AK002276.1 | - | 3, 4 | 272 | |||
AK151307.1 | - | 3, 4 | 290 | |||
AK151366.1 | - | 3, 4 | 290 | |||
AK169267.1 | - | 3, 4 | 273 | |||
BC049637.1 | - | 3, 4 | 180 | 6585654 | ||
BC057097.1 | - | 3, 4 | 269 | 5714599 |