Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3glct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081204.1 | NP_001074673.1 | 12, 13 | 1111 | ||
GenBank mRNA | AB253762.1 | - | 12, 13 | 1076 | ||
AK089440.1 | - | 12, 13 | 1111 | |||
AK145082.1 | - | 12, 13 | 1086 |