Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286663.1 | NP_001273592.1 | 5 | 1021 | ||
NM_001286664.1 | NP_001273593.1 | 5 | 1021 | |||
NM_028358.2 | NP_082634.1 | 6 | 463 | |||
NM_212468.3 | NP_997633.1 | 6 | 463 | |||
GenBank mRNA | AK013425.1 | - | 6 | 464 | ||
AK160556.1 | - | 6 | 769 | |||
BC028648.1 | - | 6 | 445 | 1314987 | ||
BC060290.1 | - | 5 | 1021 | |||
BC061183.1 | - | 6 | 444 | 30295766 |