Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164679.1 | NP_001158151.1 | 14 | 2528 | ||
GenBank mRNA | AK173222.1 | - | 12 | 3516 | ||
BC027735.1 | - | 14 | 2430 | |||
BC027790.1 | - | 7 | 2002 | |||
BC042391.1 | - | 7 | 2723 | |||
BC053460.1 | - | 2 | 567 | |||
BC059855.1 | - | 5 | 1061 |