Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164681.1 | NP_001158153.1 | 6 | 818 | ||
NM_198672.1 | NP_941074.1 | 6 | 818 | |||
GenBank mRNA | AK138932.1 | - | 6 | 817 | ||
AK149499.1 | - | 2 | 238 | |||
BC035526.1 | - | 6 | 814 | |||
BC048380.1 | - | 6 | 799 | |||
BC057187.1 | - | 6 | 803 | 5098677 | ||
BC057188.1 | - | 6 | 803 | 5100121 | ||
BC061004.1 | - | 6 | 816 | 30315941 | ||
L11333.1 | - | 6 | 793 | |||
S64130.1 | - | 6 | 794 |