Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm1110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281475.1 | NP_001268404.1 | 19, 20 | 1917 | ||
GenBank mRNA | AK136683.1 | - | 10, 11 | 1343 | ||
DQ664197.1 | - | 19, 20 | 1926 |