Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100460.1 | NP_001093930.1 | 2 | 1356 | ||
GenBank mRNA | BC023876.1 | - | 1 | 259 | ||
BC046387.1 | - | 3 | 1038 | |||
BC051084.1 | - | 1 | 1357 |