Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tvp23a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013778.1 | NP_001013800.1 | 3, 4 | 646 | ||
GenBank mRNA | BC068110.1 | - | 3, 4 | 646 | 5696118 |