Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdm14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081209.2 | NP_001074678.2 | 6 | 1271 | ||
GenBank mRNA | AK141218.1 | - | 5 | 1049 | ||
KJ463838.1 | - | 5 | 1238 |