Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145538.2 | NP_663513.1 | 4 | 405 | ||
GenBank mRNA | AK080983.1 | - | 546 | |||
AK088196.1 | - | 4 | 418 | |||
AK151326.1 | - | 391 | ||||
AK151705.1 | - | 4 | 416 | |||
AK170042.1 | - | 415 | ||||
AK213753.1 | - | 2 | 91 | |||
BC002270.1 | - | 4 | 405 | 3496088 |