Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145538.2 | NP_663513.1 | 6 | 840 | ||
GenBank mRNA | AK080983.1 | - | 981 | |||
AK088196.1 | - | 6 | 853 | |||
AK151326.1 | - | 826 | ||||
AK151705.1 | - | 6 | 850 | |||
AK199081.1 | - | 11 | ||||
BC002270.1 | - | 6 | 840 | 3496088 |