Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wfdc13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012704.1 | NP_001012722.1 | 2 | 294 | ||
GenBank mRNA | AY542486.1 | - | 2 | 294 | ||
AY542487.1 | - | 2 | 294 | |||
BC145778.1 | - | 2 | 203 | 40131081 | ||
BC145780.1 | - | 2 | 203 | 40131083 |