Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030296.4 | NP_001025467.1 | 2, 3 | 801 | ||
GenBank mRNA | AK133129.1 | - | 2, 3 | 761 | ||
BC138854.1 | - | 1, 2 | 469 | 8861876 | ||
BC138856.1 | - | 1, 2 | 469 | 8861878 |