Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Minos1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163006.2 | NP_001156478.1 | 2, 3 | 233 | ||
GenBank mRNA | AA023452.1 | - | 2, 3 | 248 | ||
AK004466.1 | - | 2, 3 | 125 | |||
AK008362.1 | - | 2, 3 | 117 | |||
AK008449.1 | - | 2, 3 | 117 | |||
AK009189.1 | - | 2, 3 | 116 | |||
AK009528.1 | - | 2, 3 | 159 | |||
AK011553.1 | - | 2, 3 | 116 | |||
AK030343.1 | - | 2, 3 | 186 | |||
AK152509.1 | - | 2, 3 | 159 | |||
AK153063.1 | - | 2, 3 | 158 | |||
AK159039.1 | - | 2, 3 | 112 | |||
AK159100.1 | - | 2, 3 | 159 | |||
AK167818.1 | - | 2, 3 | 158 | |||
AW226897.1 | - | 2, 3 | 291 | |||
BC055791.1 | - | 2, 3 | 150 | 6410794 | ||
BY776411.1 | - | 2, 3 | 223 |