Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177566.3 | NP_808234.2 | 12 | 2134 | ||
GenBank mRNA | AK052570.1 | - | 5 | 474 | ||
AK173061.1 | - | 8 | 839 | |||
BC089365.1 | - | 12 | 2134 | 6844583 | ||
BC116343.1 | - | 12 | 2110 | 40098168 | ||
BC116344.1 | - | 12 | 2110 | 40098169 |