Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005916.2 | NP_001005916.1 | 2 | 278 | ||
GenBank mRNA | AK014458.1 | - | 1 | 110 | ||
AK030262.1 | - | 2 | 167 | |||
AK154850.1 | - | 2 | 260 | |||
BC049933.1 | - | 2 | 278 | 5387663 |