Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008706.2 | NP_001008706.1 | 2 | 197 | ||
GenBank mRNA | AY314008.1 | - | 102 | |||
BC138691.1 | - | 1 | 236 | 8861713 |